Removable needles
Supplier: HAMILTON BONADUZ
207434 7758-03 7758-02 7758-04 7803-05 7803-07 7762-06 7803-04 7770-02 7803-03 7768-02 7762-01 7784-08 7780-02 7803-01
HAMI207434EA 0 SGD
HAMI207434 HAMI7758-03 HAMI7758-02 HAMI7758-04 HAMI7803-05 HAMI7803-07 HAMI7762-06 HAMI7803-04 HAMI7770-02 HAMI7803-03 HAMI7768-02 HAMI7762-01 HAMI7784-08 HAMI7780-02 HAMI7803-01
Removable needles
Syringe Needles
The benefit of a replaceable needle for the occasional bend or clog and the minimal dead volume of a cemented needle. The unique removable needle design installs the needle precisely at the zero line. The result is a zero dead volume connection that simplifies priming.
Custom lengths and point styles are available. You can order your custom made needle according to your specification.
* Large hub
** Kel-F hub
Order Now
SPECIFICATIONS
Environmentally Preferable