Anion exchange HPLC columns, RCX-30
Supplier: HAMILTON BONADUZ
79803
HAMI79803EA 5700 SGD
HAMI79803
Anion exchange HPLC columns, RCX-30
Chromatography Columns
RCX-30 carbohydrate analysis columns are designed for the isocratic or gradient separation of carbohydrates. When an isocratic method is used with a conductivity, refractive index, ultraviolet, or pulsed amperometric detector (PAD), mono and disaccharides such as glucose, fructose, sucrose and lactose can be quickly determined.
  • Greater exchange capacity than that of the PRP-X100
  • Samples with two or three carbohydrates can be quickly separated isocratically
  • More complex samples require gradient elution