Microlitre™ syringes for manual HPLC injections on Rheodyne and Valco valves
Supplier: HAMILTON BONADUZ
80365 80465 80565 80665 80765 80975 81065 81075 81216 81265
HAMI80365EA 113 SGD
HAMI80365 HAMI80465 HAMI80565 HAMI80665 HAMI80765 HAMI80975 HAMI81065 HAMI81075 HAMI81216 HAMI81265
Microlitre™ syringes for manual HPLC injections on Rheodyne and Valco valves
Syringes Syringes for Chromatography
NR = Rheodyne needle
RNR = Replacement Rheodyne needle
Order Now
SPECIFICATIONS
Model 1705 NR 1710 RNR 1710 NR 1750 NR 1750 RNR
Environmentally Preferable
Replacement needle 613-1951 613-1951 613-1951 613-1966 613-1966