Gastight® 1700 series syringes
Supplier: HAMILTON BONADUZ
80000 80001 80030 80200 80230 80900 80901 80920 80939 81000 81001 81020 81030 81100 81120 81130 81175 81217 81220 81230 7655-01 7653-01 7654-01 80085 80016 CAL81230 208639 CAL81220 81085 81185
HAMI80000EA 135 SGD
HAMI80000 HAMI80001 HAMI80030 HAMI80200 HAMI80230 HAMI80900 HAMI80901 HAMI80920 HAMI80939 HAMI81000 HAMI81001 HAMI81020 HAMI81030 HAMI81100 HAMI81120 HAMI81130 HAMI81175 HAMI81217 HAMI81220 HAMI81230 HAMI7655-01 HAMI7653-01 HAMI7654-01 HAMI80085 HAMI80016 HAMICAL81230 HAMI208639 HAMICAL81220 HAMI81085 HAMI81185
Gastight® 1700 series syringes
Syringes Syringes for Chromatography
These syringes are the Gastight® version of the original 700 series syringes and are ideal for dispensing volumes from 1 µl up to 500 µl.
  • Fluid path includes stainless steel, borosilicate glass and PTFE
  • Gas sterilisable
Gastight® syringes are ideal for dispensing both liquids and gases and have a precision machined PTFE plunger tip which creates a leak free seal. With the tight fit, the tip essentially wipes the interior of the syringe barrel free of sample. This feature is particularly useful with heterogeneous samples as it reduces the chance that aMore Product Information