Syringes for HPLC autosamplers, PerkinElmer instruments
Supplier: HAMILTON BONADUZ
81460 81360
HAMI81460EA 276 SGD
HAMI81460 HAMI81360
Syringes for HPLC autosamplers, PerkinElmer instruments
Syringes Syringes for Chromatography
Gastight syringes for use with gases or liquids. Designed with or without ChemSeal (C) termination.
  • PTFE plunger tip creates a leak-free seal
  • Syringe glass barrel and plunger are interchangeable
Cheminert (C or CX) - This termination is a 1/4"−28 fitting designed to fit directly into instrumentation. The X-style, meaning plunger stop, on this syringe is designed to limit wear and tear on the plunger tip.