Modified syringes Microliter™7000 series
Supplier: HAMILTON BONADUZ
80100 86211 88000 CAL88011
HAMI80100EA 375 SGD
HAMI80100 HAMI86211 HAMI88000 HAMICAL88011
Modified syringes Microliter™7000 series
Syringes Syringes for Chromatography
Suitable for applications such as GC, TLC and micro pipetting. Manufactured according to the principle of dead volume by positive pressure. A tungsten piston is introduced into the syringe as far as the tip of the needle. Needle and piston are calibrated against each other and must be changed at the same time in the event of replacement.
  • Volumetric dosage takes place in the needle
  • Precision ±1% of the nominal volume
Order Now
SPECIFICATIONS
Environmentally Preferable