Super syringes
Supplier: HAMILTON BONADUZ
86301
HAMI86301EA 0 SGD
HAMI86301
Super syringes
Syringes Syringes for Chromatography
Super syringes with PTFE Luer lock (TLL) connector or tracheal adapter.
  • Clear acrylic syringe barrels
  • For use with gases and liquids
Ideal for use with air sampling, preparing gas standards, calibrating reservoirs, and pneumographs.
Order Now
SPECIFICATIONS
Environmentally Preferable