Syringes, Digital Syringes™
Supplier: HAMILTON BONADUZ
DS81230
HAMIDS81230EA 6810 SGD
HAMIDS81230
Syringes, Digital Syringes™
Syringes Syringes for Chromatography
The Digital Syringe™ is a calibrated device composed of a serialised syringe and a serialised digital electronic holder.
  • Achieve precision up to 0,5%
  • Dispense volumes from 500 µl down to 50 nl
  • Device also reduces the possibility of plunger bending
A digital readout allows enhanced syringe volume readability, while an adjustable plunger stop assures accurate, reproducible sample injections.
Delivery information: Each unit is delivered with a certificate of calibration.