Syringe diluters, Microlab® 600 series
Supplier: HAMILTON BONADUZ
ML625-DIL
HAMIML625-DILEA 18980 SGD
HAMIML625-DIL
Syringe diluters, Microlab® 600 series
Dilutors Syringe Dilutors
The Microlab® 600 dual syringe diluter is a semi-automated liquid handling instrument capable of dilutions up to 1:50000 in a single step. The precision stepper motors provide exceptional positional accuracy across the range of Hamilton bubble-free prime (BFP) syringes from 10 µl to 50 ml volumes. The inert fluid path and the diluent washing of the the tubing between each sample, minimises carry-over for even the most sensitive techniques. The basic controller features a quick start and run screen for priming and programming the diluent and sample volumes, and the controller alsoMore Product Information