Stoppers with a conical ground joint
Supplier: KARTELL
1816
KART1816EA 10.4 SGD
KART1816
Stoppers with a conical ground joint
Stoppers PE/LDPE/HDPE Stoppers
PP, transparent, with 2 hose attachments, 8 mm Ø. Ideal for ventilating or transferring air, gases or liquids in a container with a normal ground joint sleeve.
  • Sealing ring for excellent sealing
  • Autoclavable
  • Octogonal holding disc for easy loosening and removal of the stopper