Holders for round bottom flasks
Supplier: KARTELL
271
KART271EA 24.1 SGD
KART271
Holders for round bottom flasks
Supports Flask Supports
Provide secure, non-slip support for round bottom flasks and any other flasks with convex bases of any height. The concentric inner rings can hold flask sizes from 10 ml to 10 L. PP, white.
  • Excellent chemical resistance
  • Thermal resistance up to +125 °C
  • Stackable