Downdraft powder station, Protector®
Supplier: LABCONCO
3410024
LBCP3410024EA 0 SGD
LBCP3410024
Downdraft powder station, Protector®
Fume Hoods and Enclosures Extraction Cabinets
Protector® Downdraft powder stations keep users safe by containing powders during fingerprint processing. The open-sided, open-top work station provides unrestricted operator movement and accommodates oversized evidence. Available in 61 and 91,4 cm widths, two or more stations may be placed side-by-side to achieve an unlimited number of width combinations. An optional transition connector mounts between two stations for a secure fit and continuous work surface.
Order Now
SPECIFICATIONS
Environmentally Preferable