Fume absorbers
Supplier: LABCONCO
3955221 3955220 3955421
LBCP3955221EA 0 SGD
LBCP3955221 LBCP3955220 LBCP3955421
Fume absorbers
Fume Hoods and Enclosures Ductless Fume Hoods
These fume adsorbers are for light duty histology cover slipping and staining areas. Activated carbon media adsorb organic odours and noxious vapors released by solvents like xylene and toluene while clean air is recirculated into the laboratory. These enclosures are low profile, completely portable and require no ductwork. They easily accommodate shallow countertops or those with overhanging cabinets.
  • Built-in, motorized impeller to provide face velocity up to 35 fpm
  • Glacier white, dry powder, epoxy-coated steel housing with perforated baffle for uniform
More Product Information