Balance enclosures, XPert™ Easy-order
Supplier: LABCONCO
3960421
LBCP3960421EA 0 SGD
LBCP3960421
Balance enclosures, XPert™ Easy-order
Fume Hoods and Enclosures Extraction Cabinets
XPert™ balance enclosures provide user protection by keeping powders, particulates and fume contained during weighing procedures. They are well suited for pharmaceutical research and other laboratory applications involving weighing of toxic substances. These vented balance safety enclosures may be ducted to the outside or connected to the FilterMate Portable Exhauster to be exhausted back into the laboratory through HEPA filters.
  • The deep interior easily accommodates large micro and analytical balances
  • Clean-Sweep air foil allows air to sweep the work
More Product Information