Gas purifiers, AtmosPure™ Re-Gen
Supplier: LABCONCO
5121835 5121831
LBCP5121835EA 20590 SGD
LBCP5121835 LBCP5121831
Gas purifiers, AtmosPure™ Re-Gen
Filters Gas Filters
The AtmosPure™ Re-Gen gas purifier produces an ultrapure inert atmosphere for materials sensitive to moisture and/or oxygen by circulating an inert gas and removing oxygen and moisture inside the box.
  • Compensation for power outages and restart of the regeneration process if interrupted for 10 minutes or longer
  • Durable epoxy coated steel housing
This purifier has the capacity to remove up to 5 litres of oxygen (13500 ppm) and 660 grams of moisture (18 million ppm) at standard temperature and pressure (STP). There are three microprocessor controlledMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Leak rating <1 x 10-6 ml/sec
Inlet/outlet ¾" compression fittings
Weight 41 kg