Freeze dryer, bench top, FreeZone®
Supplier: LABCONCO
700201050 700202050 700401050 700402050 700801050 700802050 710201050 710202050 710401050 710402050 720401050 720402050
LBCP700201050EA 0 SGD
LBCP700201050 LBCP700202050 LBCP700401050 LBCP700402050 LBCP700801050 LBCP700802050 LBCP710201050 LBCP710202050 LBCP710401050 LBCP710402050 LBCP720401050 LBCP720402050
Freeze dryer, bench top, FreeZone®
Freeze Dryers
FreeZone® bench top freeze dryers are designed for lyophilising light to moderate sample loads. The compact design is ideal for laboratories with limited floor space.
  • Consistent, high quality results with flexible programming, and on board data collection
  • Challenging samples are easy with accessories designed specifically for laboratory samples
  • Brushed stainless steel and glacier white, powder-coated steel exterior with blue accents
  • End-Zone™ end point detection option takes the guess work out of sample dryness
  • PTFE-coated collector coil and chamber for
More Product Information