Dry evaporators, RapidVap® Vertex™ Dry
Supplier: LABCONCO
7320030 7320035 7320020
LBCP7320030EA 17040 SGD
LBCP7320030 LBCP7320035 LBCP7320020
Dry evaporators, RapidVap® Vertex™ Dry
Evaporators Centrifugal Evaporators
This evaporator speeds evaporation of up to 50 samples at the same time using dry heat and nitrogen blow down. Unlike water bath heaters, the dry block heater requires less maintenance, no distilled water or additives, and adds no potential source of contamination.
  • LCD with convenient touchpad allow easy programming of time and temperature
  • Up to ten different programs may be stored in the microprocessor for protocol consistency
  • Front-mounted pressure regulator with analog display of pressure from 0 to 45 psi in 2 psi increments
  • Temperature sensor probe for
More Product Information