Scroll vacuum pump
Supplier: LABCONCO
7587300
LBCP7587300EA 0 SGD
LBCP7587300
Scroll vacuum pump
Pumps Vacuum Pumps
Scroll Vacuum Pumps are low maintenence, corrosion resistent pumps that require no oil.
  • Scrolls be changed every 40000 hours of use
  • Eliminating the oil eliminates hydrocarbon contamination
Handle water vapor better than most types of pumps and make significantly less noise during operation. Works with FreeZone triad, 1, 2, 4, 5, 6, 8, 12 and 18 L freeze dryers.