Vacuum concentrators, CentriVap® micro IR
Supplier: LABCONCO
7701030 7701035
LBCP7701030EA 13180 SGD
LBCP7701030 LBCP7701035
Vacuum concentrators, CentriVap® micro IR
Evaporators Centrifugal Evaporators
The CentriVap micro IR vacuum concentrator has a very small footprint and therefore fits in tight spaces. It is ideal for small samples such as DNA and small throughput in molecular biology, proteomics, genomics, genetics, cell biology and drug discovery labs.
  • Transparent infrared (IR) glass lid directs heat to the samples to speed evaporation and reduces cross contamination from condensation
  • Built-in PTFE-coated diaphragm pump with ultimate vacuum of 160 mBar and displacement capacity of 5,5 liters/minute
  • With micro IR strobe light to view samples while
More Product Information