Small tray dryer, FreeZone®
Supplier: LABCONCO
780701050
LBCP780701050EA 0 SGD
LBCP780701050
Small tray dryer, FreeZone®
Freeze Dryers
These FreeZone® small tray dryers provide precise control and program flexibility for consistent performance and are designed for lyophilising small to moderate bulk sample loads or numerous sample batches.
  • Precision with 3 heated shelves up to +60 °C
  • Clear acrylic viewing door provides complete visibility of the shelves
  • Temperature and vacuum level controls in 30 programs with up to 16 segments each
  • Quality reassurance with onboard data collection and real time graphing
  • Sample probes with remote notification of operating parameters or alerts