Centrifugal Concentrators, Bench Top, CentriVap®
Supplier: LABCONCO
7811030 7810033 7460035 7810030 7810031 7810032 7810034 7810035 7810036 7810037 7811035 7970030 7970035 7385030 7385035 7385037 7970037 7811037 7460037 7811036
LBCP7811030EA 7600 SGD
LBCP7811030 LBCP7810033 LBCP7460035 LBCP7810030 LBCP7810031 LBCP7810032 LBCP7810034 LBCP7810035 LBCP7810036 LBCP7810037 LBCP7811035 LBCP7970030 LBCP7970035 LBCP7385030 LBCP7385035 LBCP7385037 LBCP7970037 LBCP7811037 LBCP7460037 LBCP7811036
Centrifugal Concentrators, Bench Top, CentriVap®
Evaporators Centrifugal Evaporators
CentriVap® vacuum concentrators are available in a range of configurations. The CentriVap® DNA model is supplied with a built-in 34 litres/min diaphragm pump, two 177 ml glass traps and a DNA rotor (60×1,5/2,0 and 72×0,5 ml). Model with glass lid is recommended for evaporation of solvents that are not compatible with acrylic (acetone, acetonitrile, ethanol, DMSO, etc.). The acid resistant model is recommended for corrosive solvents (TFA, acetic acid, etc.).
  • Powder coated aluminium chamber; PTFE coated aluminium chamber with Hastelloy shaft for acid
More Product Information