FreeZone® Triad™ Freeze drying system
Supplier: LABCONCO
794001050
LBCP794001050EA 0 SGD
LBCP794001050
FreeZone® Triad™ Freeze drying system
Freeze Dryers
The FreeZone® Triad™ freeze drying system is an all inclusive bench top lyophiliser for 4 samples on the left side and stoppering tray dryer processing shelf. The processing shelf holds up to 441×3 ml serum bottles and the four valves hold samples in flasks, serum bottles or ampoules. Samples for both types of freeze drying can be run at once. The chamber pre-freezes samples to save money and time, eliminating the need for a separate freezer and product transfer.
  • Microprocessor controlled temperature programming from −55 to +50 °C during ramping and
More Product Information