Shell freezers, benchtop, FreeZone®
Supplier: LABCONCO
7949060 7949070
LBCP7949060EA 0 SGD
LBCP7949060 LBCP7949070
Shell freezers, benchtop, FreeZone®
Freezers Laboratory / General Purpose Freezers
FreeZone® benchtop shell freezers provide a compact system for prefreezing samples for lyophilization.
  • ¹/₂ hp HCFC/CFC refrigeration systems cool to −40 °C for processing samples
  • LED illuminates green, when the system reaches operating temperature
  • Right-side mounted power switch
  • Left-side mounted retractable bath drain hose
Temperature range: − 20 °C to − 42°C
Certifications: UL certified.