Cart, Flexi-Bin™
Supplier: LABCONCO
8010001
LBCP8010001EA 0 SGD
LBCP8010001
Cart, Flexi-Bin™
Trolleys
Durable and chemical-resistant, glacier white, epoxy-coated tubular steel frames. For safe transport of glassware and chemical containers.
  • Features a removable, corrosion-resistant polyethylene bin with 10" overall depth
  • Includes three long and four short polyethylene dividers for partitioning the bin to securely hold items in upright position
  • The bottom open shelf can hold additional supplies
  • With 4" diameter casters, two with toe locks
  • Ready-to-roll, full factory assembly
Order Now
SPECIFICATIONS
Environmentally Preferable