Trolleys, glasswares
Supplier: LABCONCO
8045000
LBCP8045000EA 2650 SGD
LBCP8045000
Trolleys, glasswares
Trolleys
Glassware carts, used to carry glasswares, plasticwares and small instruments in vinyl-coated wire baskets. Choose from 2-baskets or 4-basket styles complete with removable plastic drip pan to catch spills.
  • Four-inch diameter casters absorb vibration over rough, uneven floors
  • Two toe-locks keeps the cart stationary when needed
  • Benches have 5" diameter, non-marking toe-locking casters with bearings for quiet operation
Order Now
SPECIFICATIONS
Environmentally Preferable