Rubber conical gaskets Guko, DURAN®
Supplier: DWK Life Sciences
292020001 292022705 292022302 292023204 292023607 292023907 292024303 292021203 292021709
SCOT292020001EA 106 SGD
SCOT292020001 SCOT292022705 SCOT292022302 SCOT292023204 SCOT292023607 SCOT292023907 SCOT292024303 SCOT292021203 SCOT292021709
Rubber conical gaskets Guko, DURAN®
Supports Support Sleeves
EPDM rubber.
Conical gaskets to place between filter funnel and filter flask.
Order Now
SPECIFICATIONS
Environmentally Preferable