Conical sleeves for vacuum filtration, Sterilin®
Supplier: Sterilin
CON0006
SLINCON0006EA 0 SGD
SLINCON0006
Conical sleeves for vacuum filtration, Sterilin®
Supports Support Sleeves
Red natural rubber cones for use with filtering flasks.
  • Set of six pieces
  • Set includes each one of 7/15 mm, 10/22 mm, 15/28 mm, 16/36 mm, 22/42 mm and 26/52 mm (inner diamater at low end/inner diameter at top end)