VWR®, Stoppers, PE
Supplier: VWR Collection
VWRI212-0550EA 27.6 SGD
VWRI212-0550 VWRI212-0551 VWRI212-0552 VWRI212-0553 VWRI212-0554 VWRI212-0572 VWRI212-0573 VWRI212-0574 VWRI212-0575 VWRI212-0576 VWRI212-0577 VWRI212-0578 VWRI212-0664 VWRI212-0665 VWRI212-0666 VWRI212-0667 VWRI212-0679
VWR®, Stoppers, PE
Stoppers PE/LDPE/HDPE Stoppers
Stopper for test tubes Ø 12 to 16 mm, polyethylene, with tongues.
  • Choice of colours