VWR®, Stoppers for Standard Ground Joints
Supplier: VWR Collection
VWRI217-0210EA 19.1 SGD
VWRI217-0210 VWRI217-0211 VWRI217-0212 VWRI217-0213 VWRI217-0214 VWRI217-0215
VWR®, Stoppers for Standard Ground Joints
Stoppers PE/LDPE/HDPE Stoppers
Greaseless closure, which prevents ground joints from seizing. With inclined, circumferential sealing lips for better sealing and reduced risk of breakage. Easily detachable. With ergonomically shaped grip.
  • PTFE, solid
  • Excellent chemical resistance
  • Gas, liquid and vacuum-tight
  • Temperature resistant from –200 to +300 °C
Order Now
SPECIFICATIONS
Environmentally Preferable