Stopper, PTFE, key-hole, color-coded, PYREX®
Supplier: Corning
7630-13 7630-16 7630-19 7630-22 7630-9
CORN7630-13EA 87.5 SGD
CORN7630-13 CORN7630-16 CORN7630-19 CORN7630-22 CORN7630-9
Stopper, PTFE, key-hole, color-coded, PYREX®
Stoppers PE/LDPE/HDPE Stoppers
PTFE. Ideal for flasks that are grounded to [ST] stopper dimensions.
  • The colour-coded keys make sorting and selecting the correct size faster and easier