Rotary evaporator traps, PYREX®
Supplier: Corning
4000-10019 4000-10024 4000-1L24 4000-25014 4000-25024
CORN4000-10019EA 216 SGD
CORN4000-10019 CORN4000-10024 CORN4000-1L24 CORN4000-25014 CORN4000-25024
Rotary evaporator traps, PYREX®
Vacuum Traps
These PYREX® traps are used in rotary evaporators. They have a round body with upper and lower stems.
  • Lower stem has a Standard Taper inner joint at the bottom and extends upward inside the body
  • The closed top of this stem has two side openings
  • The upper stem has a Standard Taper outer joint