PESTINORM® headspace solvents
High purity solvents for organic volatile impurity analysis
Static headspace GC (GC-HS) is a technique used to concentrate volatile analytes prior to analysis. It can improve detection of low levels of volatile analytes and minimise matrix interference by eliminating the need to inject the sample directly. Our new GC-HS solvent line includes six of the most commonly used organic solvents:
- Very high purity (>99,99% of assay)
- Very high UV transmittances
- Each batch is controlled and specifically evaluated by analysis against OVI standards
- Suitable for the analysis of OVI as described by the ICH, USP and EP methods
Log in to see your contract pricing and availability.