Masterflex Flexible Tubing
Flexible Tubing... Even Under Pressure!
Pump at pressures up to 100 psi with peristaltic pump tubing? It's now a reality with our NEW Masterflex® High-Pressure Norprene® tubing.
Choose from two sizes: L/S® 16HP tubing (MFLX06504-16), which delivers 100 psi maximum continuous pressure and 125 psi intermittent pressure, with a flow range of 0.9 to 90 mL/min. L/S® 15HP tubing (MFLX06504-15) produces a flow range of 1.7 to 170 mL/min, with 80 psi maximum continuous pressure and 100 psi maximum intermittent pressure. Besides higher pressure, this innovative tubing provides excellent flexibility, so it can be easily used in a wide variety of pump applications.
Masterflex® High-Pressure Norprene® tubing is designed for the Masterflex® L/S® high-pressure tubing pump system (MFLX77914-10) , which transfers fluids at continuous pressures up to 100 psi, with a flow range of 0.09 to 170 mL.min.
Continuous pressure up to 100 psi is possible with High-Pressure Norprene® MFLX06504-15 tubing.
Typical Applications
- Cosmetic production
- Pharmaceutical
- Biotechnology
- Water treatment
- Food and Environmental
- Or other high-pressure applications!