Liquid Pumps Technical Equations and Information Tables
Efficiency Formulas for Rotary Positive Displacement Pumps
Pipe Velocity
- It is best to allow 10 pipe diameter upstream and 5 pipe diameter downstream leading to the pump suction.
- Try to keep pipe velocities around 10 ft/sec for good practical results.
- Doubling the pipe diameter increases the capacity of the pipe four times.
Rules of Thumb for Motors
At 3600 rpm: motor develops 1.5 ft/lb per hp
At 1800 rpm: motor develops 3.0 ft/lb per hp
At 1200 rpm: motor develops 4.5 ft/lb per hp
At 575 volts, a 3-phase motor draws 1.00 amps per hp
At 460 volts, a 3-phase motor draws 1.25 amps per hp
At 230 volts, a 3-phase motor draws 2.50 amps per hp
Approximate rpm at Full Load for Medium Sized Motors
Poles | rpm 60 Hz | Synch speed | rpm 50 Hz | Synch speed |
---|---|---|---|---|
2 | 3500 | 3600 | 2850 | 3000 |
4 | 1750 | 1800 | 1450 | 1500 |
6 | 1150 | 1200 | 950 | 1000 |
8 | 850 | 900 | 700 | 750 |
Loss of Air Pressure Due to Friction
(Per 100 Feet of Pipe, 100 psi Initial Pressure)
Free-air capacity in cfm (at 14.7 psi) |
Equivalent capacity in cfm (at 100 + 14.7 psi) |
1/2" | 3/4" | 1" | 1-1/4" | 1-1/2" | 2" |
---|---|---|---|---|---|---|---|
10 | 1.28 | 1.38 | 0.09 | 0.03 | 0.007 | ||
20 | 2.56 | 1.42 | 0.34 | 0.10 | 0.026 | 0.012 | |
30 | 3.84 | 3.13 | 0.74 | 0.23 | 0.056 | 0.026 | |
40 | 5.13 | 5.55 | 1.28 | 0.38 | 0.096 | 0.044 | 0.013 |
50 | 6.41 | 8.65 | 2.00 | 0.60 | 0.146 | 0.067 | 0.020 |
60 | 2.84 | 0.84 | 0.210 | 0.095 | 0.027 | ||
70 | 3.85 | 1.12 | 0.280 | 0.130 | 0.036 | ||
80 | 5.01 | 1.44 | 0.360 | 0.160 | 0.046 | ||
90 | 6.40 | 1.85 | 0.450 | 0.200 | 0.058 | ||
100 | 7.80 | 2.21 | 0.550 | 0.250 | 0.069 | ||
125 | 12.40 | 3.41 | 0.850 | 0.380 | 0.107 | ||
150 | 18.10 | 4.91 | 1.200 | 0.540 | 0.150 | ||
175 | 6.80 | 1.640 | 0.730 | 0.200 | |||
200 | 8.79 | 2.120 | 0.950 | 0.260 | |||
250 | 3.300 | 1.480 | 0.400 | ||||
300 | 4.710 | 2.100 | 0.570 | ||||
350 | 6.450 | 2.860 | 0.770 | ||||
400 | 8.300 | 3.700 | 0.990 | ||||
450 | 4.650 | 1.270 | |||||
500 | 5.790 | 1.560 | |||||
600 | 8.45 | 2.230 | |||||
700 | 3.000 | ||||||
800 | 4.000 | ||||||
900 | 5.050 | ||||||
1000 | 6.200 |
Loss of Air Pressure Due to Pipe Bends
(Per 100 Feet of Straight Pipe)
Angle of pipe bend | Loss of pressure in psi per pipe ID | |||||
1/2" | 3/4" | 1" | 1-1/4" | 1-1/2" | 2" | |
45° | 0.73 | 0.92 | 1.18 | 1.55 | 1.85 | 2.35 |
90° | 1.60 | 2.00 | 2.50 | 3.40 | 4.00 | 5.10 |
Flow of Air through Orifice
(with Discharge of Orifice at 14.7 psia and 70°F)
Supply pressure (psig) |
Flow of air in cfm per orifice size | |||||||||
1/32" | 1/16" | 3/32" | 1/8" | 5/32" | 3/16" | 7/32" | 1/4" | 9/32" | 5/16" | |
65 | 1.15 | 4.49 | 10.10 | 17.90 | 27.90 | 40.30 | 55.20 | 71.80 | 89.90 | 111.70 |
70 | 1.21 | 4.77 | 10.80 | 19.10 | 29.70 | 42.80 | 58.80 | 76.40 | 95.70 | 118.80 |
75 | 1.30 | 5.06 | 11.40 | 20.20 | 31.50 | 45.40 | 62.30 | 81.00 | 105.50 | 126.00 |
80 | 1.37 | 5.35 | 12.10 | 21.10 | 33.30 | 48.00 | 65.80 | 85.60 | 107.40 | 133.10 |
85 | 1.44 | 5.64 | 12.70 | 22.50 | 35.10 | 50.60 | 69.40 | 90.30 | 113.20 | 140.30 |
90 | 1.52 | 5.92 | 13.40 | 23.70 | 36.90 | 53.20 | 72.90 | 94.80 | 119.00 | 147.50 |
95 | 1.59 | 6.21 | 14.00 | 24.80 | 38.70 | 55.70 | 76.50 | 99.40 | 124.90 | 154.60 |
100 | 1.66 | 6.50 | 14.70 | 26.00 | 40.50 | 58.30 | 80.00 | 104.60 | 130.70 | 161.80 |
125 | 2.03 | 7.94 | 17.90 | 31.70 | 49.50 | 71.40 | 97.78 | 127.10 | 159.80 | 197.50 |
150 | 2.40 | 9.28 | 21.20 | 37.50 | 58.40 | 84.40 | 115.40 | 150.40 | 189.00 | 233.30 |