NORPRENE® (A 60 G) Tubing
Advantages:
- Best choice for vacuum/pressure applications.
- Offers longest life with good flow consistency.
- Heat and ambient ozone resistant.
- Good resistance to acids and alkalies.
- Black color hides dirt and dust.
- Heat sealable, nonaging, and nonoxidizing.
- High dielectric constant.
Limitations:
- Potential leaching of USP mineral oil or blend material.
Application Suitability:
Acids: | Good |
---|---|
Alkalies: | Good |
Organic solvents: | Not recommended |
Pressure: | Excellent |
Vacuum: | Excellent |
Viscous fluids: | Excellent |
Sterile fluids: | Not recommended |
Physical Characteristics and Compostion:
- Thermoplastic elastomer.
- Polypropylene-based material with USP mineral oil.
- Excellent tensile strength.
- Firm (stiff) material.
- Opaque, black.
Temperature Range:
-75 to 275°F (-59 to 135°C)
Meets Classifications:
- None
Gas Permeability:
cc x mm
x 10-10
(cm2 x sec. x cm Hg
CO2: | 1200 |
---|---|
H2: | - |
O2: | 200 |
N2: | 80 |
Cleaning/Sterilization:
- Sterilization is not recommended.