Masterflex I/P Tubing Options
Achieve top performance from your Masterflex I/P pump heads when you use precision-extruded tubing. Tubing is manufactured to tight tolerances that match our I/P pump heads. Choose from precision tubing or high-performance precision tubing. High-performance precision tubing has a thicker wall that improves pressure generation, suction lift, ability to pump viscous fluids, and tubing life. Choose from over a dozen different formulations.
- BioPharm Plus Silicone (platinum-cured)
- Pharmed®
- Tygon LFL
- Silicone (platinum-cured)
- Silicone (peroxide cured)
- C-Flex
- Norprene food
- BioPharm Silicone (platinum-cured)
- Tygon Food
- Tygon lab
- Norprene
- Tygon Fuel and Lubricant
- Viton®
- FDA compliant Viton®
Precision Tubing Specifications
Tubing size | I/P 26 | I/P 73 | I/P 82 | |
---|---|---|---|---|
Inside diameter (nominal) | 0.25" (6.4 mm) | 0.37" (9.5 mm) | 0.5" (12.7 mm) | |
Hose barb size (nominal) | 1/4" (6.4 mm) | 3/8" (9.5 mm) | 1/2" (12.7 mm) | |
Flow range (approximate) with 20 to 650 rpm drive | 0.12 to 4 L/min (0.03 to 1.1 GPM) |
0.2 to 8 L/min (0.05 to 2.1 GPM) |
0.4 to 13 L/min (0.1 to 3.5 GPM) |
|
Maximum pressure, continuous** | 25 psig (1.7 bar) | 10 psig (0.7 bar) | ||
Maximum pressure, intermittent** | 40 psig (2.7 bar) | 20 psig (1.4 bar) | ||
Maximum vacuum** | 26" Hg (660 mm Hg) | 20" Hg (510 mm Hg) | ||
Suction lift | 29 ft H2O (8.8 m H2O) | 23 ft H2O (7.0 m H2O) |
High-Performance Precision Tubing Specifications
Tubing size | I/P 70 | I/P 88 | I/P 89 | |
---|---|---|---|---|
Inside diameter (nominal) | 0.37" (6.4 mm) | 0.5" (12.7 mm) | 0.62" (15.88 mm) |
|
Hose barb size (nominal) | 3/8" (9.5 mm) | 1/2" (12.7 mm) | 5/8" (15.88 mm) | |
Flow range (approximate) with 20 to 650 rpm drive | 0.24 to 8 L/min (0.06 to 10 GPM) |
0.5 to 15 L/min (0.05 to 2.1 GPM) |
0.52 to 17 L/min (0.14 to 4.5 GPM) |
|
Maximum pressure, continuous** | 25 psig (1.7 bar) | 20 psig (1.4 bar) | 15 psig (1.0 bar) | |
Maximum pressure, intermittent** | 40 psig (2.7 bar) | 35 psig (1.4 bar) | 25 psig (1.4 bar) | |
Maximum vacuum** | 26" Hg (660 mm Hg) | 24" Hg (610 mm Hg) | ||
Suction lift | 29 ft H2O (8.8 m H2O) | 27 ft H2O (8.2 m H2O) |
**Actual performance varies depending on tubing formulation—values shown are for firm tubing.