KC Kimtech G6 Cleanroom Gloves
New Products Launch in Our Region. Your Ideal Hands Protection Armour in various sizes!
Ambidextrous cleanroom gloves feature textured fingertips for precision work in pharmaceutical, biotech, semiconductor, medical device, data storage, and touch panel applications.
Ideal for use in pharma grade C&D, ISO 6 - 8, and Class 1000 cleanrooms.
Click on "Request a Quote" to enjoy our exclusive deals now!
- Non-sterile, powder-free nitrile gloves
- AQL 1,5
- 5,1 mil/0,13mm thickness at fingertips
- Double-bagged for cleanroom use
- Accelerator-, DOP-, and amide-free
- Silicone-free material
- Beaded cuff for easy donning
Description | UOM | VWR Catalog Number |
KIMTECH G6 WHITE NITRILE GLOVE, XS SIZE, 290mm | 1*1,000 items | KIMB68700 |
KIMTECH G6 WHITE NITRILE GLOVE, S SIZE, 290mm | 1*1,000 items | KIMB68701 |
KIMTECH G6 WHITE NITRILE GLOVE, M SIZE, 290mm | 1*1,000 items | KIMB68702 |
KIMTECH G6 WHITE NITRILE GLOVE, L SIZE, 290mm | 1*1,000 items | KIMB68703 |
KIMTECH G6 WHITE NITRILE GLOVE, XL SIZE, 290mm | 1*1,000 items | KIMB68704 |