IKA RCT Heat Safely
Integrated Safety protects both sample and user
Since every sample is different, it is possible to individually set the maximum temperature allowed to ensure its safety. Integrated Safety from IKA ensures that the respective flashpoint of the medium used is not reached, independently of any device defects, glass breakages, or physical effects such as thermal distortion.
Click on "Request a Quote" to enjoy our exclusive deals now!
- Independent control circuits - In addition to the sensors which directly measure the sample and the hot plate, Integrated Safety also provides an additional, absolutely independent, safety circuit on the hot plate.
- Safety glass. - The latest generation of our magnetic stirrers have a display with hardened, unbreakable glass which is also particularly resistant to chemicals.
- Locking function. - A locking function prevents accidental changes to the set speed and temperature values. Safety circuits are also adjustable.
Description | UOM | VWR Catalog Number |
Magnetic stirrer with heating incl. RCT basic, PT 1000 temp. sensor, & fixing device | 1*1 items | IKAA10000294 |