PAN Biotech Viral Transport Medium
A ready to use medium for transport of viral samples, such as throat and nasal swabs taken from Covid- 19 suspects..
Viral Transport Media (VTM) are necessary for the transport of nasopharygeal /oropharygeal swabs. PAN VTM HBSS includes antibiotics and antimycotics to supress bacterial and fungal contamination. PAN VTM HBSS contains phenol red as pH indicator to ensure medium integrity at time of specimen collection.
Click on "Request a Quote" to enjoy our exclusive deals now!
- Proven and validated viral transport medium (validation includes human corona virus)
- Formulation similar to Copan Medium (WHO recommended)
- Flexibility in transport temperature: from 2…25°C
Request a Quote
Description | UOM | VWR Catalog Number |
HBSS PAN VTM 3X50ML | 1 * 150 ml | PANBP04-01753 |
HBSS PAN VTM | 1 * 1,000 ml | PANBP04-01754 |
HBSS PAN VTM Kit | 1 * 1 kit | PANBP04-01753K |