NanoEntek Cell Counting C-Chip
Hurry! Year End Grand Sale. New! Count your cells safely (No glass). Enjoy exclusive prices when you purchase 2 boxes or more.
Quartz grade optical plastic counting chambers applicable in blood analysis (hematology).
Suitable for cell counting and culture, concentration measurement and viability test, in bacteria and fungal spore counting, IVF, IUI tasks of sperm counting.
Click on "Request a Quote" to enjoy our exclusive deals now!
- Guaranteed repeatability and reliability, eliminate the need for washing and reuse
- Disposable hemocytometers that do not need coverslips
- Chip to reduce exposure to infectious samples and contamination
View Cell Counter ADAM-MC
.
Description | UOM | VWR Catalog Number |
HEMOCYTOMETER C-CHIP NEUBAUER 100μM | 1 * 50 items | NANODHC-N01 |
HEMOCYTOMETER C-CHIP 4CH NEUBAUER 100μM | 1 * 50 items | NANODHC-N04 |