VWR ULT Freezers
New Products Launch in Our Region. Unbelievable Prices!
These upright freezers have been designed for sample protection and ease of use.
All upright freezers feature a convenient, eye-level control and monitoring system.
Features heavy gauge, cold-rolled steel cabinet construction with a powder
coated paint finish, resistant to chipping and rust. Four inner doors reduce cold
air loss and improve temperature recovery after door openings. Sturdy, stainless
steel shelving (adjustable in 25 mm increments). Single-handed operation with
ergonomic, padlock-compatible door handle featuring an integrated key lock.
Click on "Request a Quote" to enjoy our exclusive deals now!
- Reduced/low peak variation to protect sample temperature
- Lower energy consumption and heat emission cost with low noise level
- Fast recovery from opening door
- Insulation: High density water blown PU foam with Vacuum Insulation Panel (VIP)
- Gaskets maximise cabinet temperature and work to eliminate frost build up
Description | UOM | VWR Catalog Number |
ULT freezer, 422 l | 1 * 1 Item | VWRI471-1228 |
ULT freezer, 549 l | 1 * 1 Item | VWRI471-1229 |
ULT freezer, 682 l | 1 * 1 Item | VWRI471-1230 |
ULT freezer, 816 l | 1 * 1 Item | VWRI471-1231 |