Eppendorf Concentrator Plus
Concentrator Plus that allows fast and carefree vacuum concentration of your samples.
This ergonomic, intuitive concentrator provides quick, efficient and gentle vacuum concentration of DNA/RNA, nucleotides, proteins and other liquid or wet samples. The coated lid provides superior chemical resistance against aggressive acids
and organic solvents (e.g. TFA, DMSO).
Comes in 3 system configurations: Basic system, Complete system with pump, and Complete system with vacuum port.
Click on "Request a Quote" to enjoy our exclusive deals now!
*Eppendorf promotion is applicable to Singapore based customers only
- Chemical resistant, maintenance-free PTFE diaphragm pump eliminates the need for changing pump oil
- Very quiet operation (<50 dB(A)), small footprint
- Simple operation due to intuitive control panel
Description | UOM | VWR Catalog Number |
Concentrator plus, complete system with integral diaphragm vacuum pump and 48×1,5/2,0 ml fixed-angle rotor, UK-plug | 1*1 items | EPPE5305000.363 |