Pan Biotech ISF-1 Medium
Culturing hybridoma cells for Antibody purification. Buy 10 of the same model and Get *1 of
the same model FREE! T&Cs Apply!
ISF-1 is a serum-free, chemically defined medium for culture of hybridoma cell lines and the production of monoclonal Antibodys (mAb).
Because of the known composition of serum-free media there is less batch to batch variability, which leads to a better comparability of different experiments. Also the risk of viral or mycoplasma contamination is very low. MAbs produced in serum-free medium are easier to purify, which safes time in the downstream processing. ISF-1 supports cell growth and antibody production without the addition of growth factors.
Compared to other serum-free media ISF-1 is superior concerning cell growth, viability and productivity.
Click on "Request a Quote" to enjoy our exclusive deals now!
- Store at 2 to 8 °C
- Sterile
*All free gifts promotions are subjected to Terms & Conditions and may change without prior notice.
Description | UOM | VWR Catalog Number |
ISF-1 medium for hybridoma, with stable glutamine, insulin human recombinant and 2,438 g/L NaHCO₃ | 1 * 1L | PANBP04-995968 |