NEWB Innova 42R Incubator Shaker with Refrigeration
Benchtop or floor mounted stackable orbital shakers that utilize a triple eccentric counter balanced drive mechanism.
These incubating orbital shakers are available with and without refrigeration, R denotes refrigeration. Designed to maximise valuable laboratory space the units can be positioned on or under bench or stacked up to two units high with optional stacking kits. The program controller is easy to use and the display, positioned at the top of the unit, can be easily seen from all angles. Intuitive programming allows multiple parameter changes to speed, optional photosynthetic lights and optional UV light decontamination on a timed basis (R models only).
There is a single height adjustable shelf for storage of incubating plates or other samples. The moisture proof, in-chamber power socket provides power for an additional instrument for example a rocker, roller or a second shaker. There is an interior light to assist viewing and the viewing window with double pane thermal glass gives visibility and insulation. These versatile shakers can accommodate flasks up to 6 litres, an RS232 serial port provides two-way communication for data logging and unit control.
Click on "Request a Quote" to enjoy our exclusive deals now!
- Large easy to read displays shows set point and actual values
- Integral timer, 0,1 to 99,9 h, shows elapsed time or counts down, automatic switch-off and audible alarm when program is complete
- Microprocessor controller maintains precise temperature, stores programs for repeat use and recalls last set points, even when unit has been shut off. Will also restart if power is interrupted
- Triple eccentric counterbalanced drive with brushless motor ensures uniform motion regardless of where the sample is on the platform even when fully loaded and operating at top speed
- Air circulation system ensures rapid temperature equilibration and uniformity, multi-function reservoir humidifies the chamber to minimise sample evaporation
Ordering information: Supplied without platforms or accessories, required accessories must be ordered separately. Factory fitted options such as 12-port gassing manifold, culture drawer, humidity probe, stacking kits, ultraviolet germicidal light or programmable photosynthetic light bank for the 42R are available, please contact your local VWR office before placing your order as options can not be retrofitted.
Description | UOM | VWR Catalog Number |
Innova 42R Stackable, On-or-Under-Bench Incubator Shaker with Refrigeration, 1 in (2.5 cm) Orbit, 230V, 50Hz (M1335-0016) + Universal Platform 46 × 46 cm (M1250-9902) | 1*1 items | NEWBM1335-0316 |