All Purpose Paraflim ® M
Special Promotion while stocks last!
An All Purpose Laboratory Film
Protect the integrity of your laboratory and research applications by keeping moisture loss to a minimum and shield the contents of your items with a flexible covering and overwrap.
- Prevents moisture and volume loss due to material composition and seal strength
- Self-sealing and pliability allows it to cling around irregular shapes and surfaces
- Stretches to more than 200% of its original length
Description | UOM | VWR Catalog Number |
---|---|---|
Parafilm for packaging, semi-transparent thermoplastic Roll, 51 mm×76,2 m, thickness: 0,127 mm |
1*1 Roll | BEMCPM992 |
Parafilm for packaging, semi-transparent, thermoplastic Roll, 102 mm×38,1 m, thickness: 0,127 mm |
1*1 Roll | BEMCPM996 |