Panbiotech Pancoll Cell Separations
Cell separation solutions for your gene expression studies and diagnostic procedures
Buy 10 of the same model and Get *1 of the same model FREE! T&Cs apply! Eg: Buy PANBP04-60100 x 10 qty and Get PANBP04-60100 x 1 qty FREE!
The isolation of cells is the first step for gene expression studies or in diagnostic procedures. Besides biological separation techniques physical separation methods are most commonly used. These methods use physical differences such as size and weight of the particles to be separated. For this purpose
so-called separating solutions (= centrifugation media) are used.
Pancoll separating solutions contain a polysaccharide with a molecular weight of 400000 daltons; this hydrophilic polymer allows for production of aqueous solutions for cell separation with a density of up to 1,2 g/ml.
PAN-Biotech offers a variety of ready-to-use products with a density of 1,063 g/ml up to 1,091 g/ml for a very wide range of cell separation applications.
Click on "Request a Quote" to enjoy our exclusive deals now!
- When properly stored, separating solutions are stable for at least 36 months
- The storage period starts with the manufacturing date
- Steriled
- Store at +2 °C to room temperature
*All free gifts promotions are subjected to Terms & Conditions and may change without prior notice.
Description | UOM | VWR Catalog Number |
Pancoll human, density/Dichte 1,077 g/ml | 1 * 100 ml | PANBP04-60100 |
Pancoll human, density 1,077 g/ml (50ml) | 1 * 25 items | PANBP04-60125 |
Pancoll human, density/dichte 1,077 g/l | 1 * 500 ml | PANBP04-60500 |
Pancoll monocytes, density/Dichte 1,068 g/ml | 1 * 100 ml | PANBP04-68100 |
Pancoll monocytes, density/Dichte 1,068 g/ml | 1 * 500 ml | PANBP04-68500 |