JT Baker and VWR Syringe Filters
New Products Launched. Up to 29% OFF! While stocks last. *T&Cs. Apply!
JT Baker Syring Filters for HPLC
These syringe filters are specifically designed for chromatography sample preparation
applications and optimised to provide the most consistent results with less extractables.
- HPLC certified
- Biosafe according to Class VI plastics tests
- Polypropylene housing material
VWR Syring Filters
Typical applications are clarification, sterile filtration, sample preparation and sterile venting.
- Sterile blister packed filters are ideal for sterile filtration
- Non sterile syringe filters are excellent for HPLC applications
- Biosafe is according to Class VI plastics tests
Certifications: Manufactured under ISO 9001:2015 quality management.
Available with nylon, PES, PTFE, PVDF, CA, RC membrane, and GF (Glass Fibre), VWR range syringe filters are idea for small volume filtration, sample preparation and sterile venting.
CA (cellulose acetate) or PES (polyether sulfone) membranes in VWR range of syringe filters are ideal for sterile filtration of buffers, tissue culture media or additives, and other biologicals. The PES membrane offers high flow rates. RC and PTFE hydrophilic type in this range offer the best chemical resistance for both aqueous solutions and solvents.
J.T. Baker range syringe filters are specifically designed for chromatography sample preparation applications and optimised to provide the most consistent results with less extractables.
Made from polypropylene housing material, they are HPLC certified, and biosafe according to Class VI plastics tests.
J.T. Baker range syringe filters are available with PTFE (hydrophilic or hydrophobic), nylon, PES, RC membrane and GF (glass fibre).
Both ranges of syringe filters are manufactured under ISO 9001:2015 quality management.
Description | UOM | VWR Catalog Number |
J.T.BAKER SF H-PTFE 13MM 0.2M NS | 1 * 200 items | BAKRSF01-14 |
J.T.BAKER SF H-PTFE 25MM 0.2M NS | 1 * 200 items | BAKRSF01-22 |
J.T.BAKER SF NYLON 25MM 0.2M NS | 1 * 200 items | BAKRSF01-50 |
J.T.BAKER SF NYLON 25MM 0.45M NS | 1 * 200 items | BAKRSF01-54 |
J.T.BAKER SF PES 25MM 0.2M NS | 1 * 200 items | BAKRSF01-98 |
J.T.BAKER SF PES 25MM 0.45M NS | 1 * 200 items | BAKRSF02-04 |
J.T.BAKER SF RC 25MM 0.2M NS | 1 * 200 items | BAKRSF02-76 |
J.T.BAKER SF RC 25MM 0.45M NS | 1 * 200 items | BAKRSF02-80 |
SYRINGE FILTER PES 25MM 0.22 MICRON NS | 1 * 100 items | VWRI514-1264 |
SYRINGE FILTER PES 25MM 0.22 MICRON ST | 1 * 50 items | VWRI514-1263 |
SYRINGE FILTER PTFE 25MM 0.45 MICRON NS | 1 * 100 items | VWRI514-1253 |
SYRINGE FILTER NY 30MM 0.22 MICRON ST | 1 * 50 items | VWRI514-1266 |
SYRINGE FILTER NY 25MM 0.22 MICRON NS | 1 * 100 items | VWRI514-1268 |
SYRINGE FILTER PVDF 30MM 0.22 MICRON NS | 1 * 100 items | VWRI514-1248 |
SYRINGE FILTER PVDF 30MM 0.22 MICRON ST | 1 * 50 items | VWRI514-1247 |
*Terms and Conditions:
- Promotion valid from now till 31 Dec 2022 *While Stocks Last.
- VWR standard Terms and Conditions apply