QuantaBio Extracta Plus DNA and RNA Kits
NEW Arrival! Nucleic Acid Purification Kits: Get your free sample today!
Buy 3, Get 1 FREE! *T&Cs Apply!
Extracta Plus DNA kits
Rapid extraction and purification of high-quality total DNA from cultured cells or tissue
- High yield – Obtain large quantities of high quality DNA, free from PCR inhibitors
- Versatility in sample types – Rapid purification of fresh or frozen tissue, cells, blood or bacteria
- Lab friendly – Silica based column eliminates toxic organic chemicals such as phenol/chloroform
- Downstream compatibility – Ideal for sensitive applications such as endpoint PCR, qPCR and next generation sequencing
Extracta Plus DNA kits offer rapid purification of total DNA from a variety of sample sources. Genomic, mitochondrial and pathogenic DNA can be collected from fresh/frozen animal cells, tissue, blood, and bacteria. The Extracta Plus Spin Column workflow enables simultaneous processing of multiple samples into purified DNA suitable for PCR, Southern blotting, RAPD, AFLP and next generation sequencing applications.
Extracta Plus RNA kits
Rapid extraction and purification of high-quality total RNA from cultured cells or tissue
- Rapid protocol – from cells or tissue samples to high-quality total RNA in just 25 minutes
- Streamlined gDNA removal – novel DNA removal column eliminates gDNA contamination and need for DNase treatment
- Extraction of low input quantities – ability to extract RNA from as little as 10 cells
- Downstream compatibility – ideal for applications such as RT-qPCR and next generation sequencing
The Extracta Plus RNA kit provides rapid extraction and purification of total RNA from cultured human or animal cells and from tissue samples. The Extracta Plus spin column workflow enables simultaneous processing of multiple samples in 25 mins to yield purified total RNA suitable for a range of downstream applications. The kit includes specially designed Extracta Plus DNA removal columns for effective removal of genomic DNA contamination, providing a convenient solution for applications that are sensitive to low amounts of DNA contamination such as RT-qPCR and NGS
Description | UOM | VWR Catalog Number |
EXTRACTA PLUS DNA 10 RXN | 1*1 Kit | QUNT95213-010 |
EXTRACTA PLUS DNA 50 RXN | 1*1 Kit | QUNT95213-050 |
EXTRACTA PLUS RNA 10 RXN | 1*1 Kit | QUNT95214-010 |
EXTRACTA PLUS RNA 50 RXN | 1*1 Kit | QUNT95214-050 |
*Terms and Conditions:
- Promotion valid from now till 30th June 2022
- Buy 3 Get 1 Free promotion is only for products of the same catalog number in 1 purchase order
- VWR standard Terms and Conditions apply