Pesticide Testing Chemicals and Columns
Avantor offers solutions to your chemical analysis in pesticide testing from gas chromatography to liquid chromatography
Avantor™ products support environmental laboratory applications for the detection and quantitation of organic parameters in soil, water, waste and other environmental samples. A pesticide residue analysis identifies the pesticide residue levels in food products by chemical analysis and gives food manufacturers and consumers’ confidence of their product ingredients.
From VWR Chemicals, VWR offers PESTINORM® solvent range that are guaranteed specifically for use in pesticide residue analysis. The range offers extremely low halogenated and other organic compounds to prevent sample contamination during sample preparation. VWR’s PESTINORM® SUPRATRACE solvents range is the universal solvent range that allows you to test your samples irrespective of sample matrix (e.g. water or soil) or detection methods (GC-ECD, GC-FID, GC-MS).
- Download VWR PESTINORM® brochure for product information.
The J.T.Baker® high purity solvents and reagents will give you the performance you need with minimal risk of contaminants that can limit accuracy and productivity and maximize the sensitivity and detecting power of your instrumentation. Improve the speed and reproducibility of your analysis with BAKERBOND Speedisk™ SPE columns and disk and Hichrom chromatography columns from Avantor ™.
- Download Avantor Guide for Cannabis testing for application notes.
We have also selected a range of high performance, sensitive and reproducible columns for your pesticide analysis below.
Products Selection | Description | Type | |
Chemicals | |||
VWR Pestinom | VWR Pestinom | Solvents and Standards | View |
Honeywell Chromasolv | Honeywell Chromasolv | Solvents | View |
Hichrom Columns | |||
HIGCHI06-10-010-5 | General purpose low-to-mid-polarity phase for a wide range of applications, including (for example): Aroclors, pesticides, herbicides and more. | GC | View |
HIGCHI05-10-010-5 | General purpose column for solvent impurities, PCBs, hydrocarbons, essential oils, semivolatiles, allergens, pesticides, etc. | GC | View |
HIGCHI55-32-025-30 | Application specifc columns developed for pesticides, herbicides, insecticides analysis etc. | GC | View |
HIGCHI06-25-030-30 | High quality. Fully Validated. | GC | View |
HIGCHI02-32-025-30 | Really versatile general purpose low-polarity phase for a wide range of applications, including (for example): PCBs, essential oils, pesticides, semivolatiles and many more. | GC | View |
HIGCHI92-32-025-30 | High Temperature general purpose column. Ideal for confrmation analysis, ideal as mid-polar column in GCxGC-HT confgurations. | GC | View |
ACETCORE-25A-1002U | Avantor® ACE® UltraCore™ SuperC18™ solid core (core shell) particle columns are ideal for demanding MS work offering improved MS signal intensity and maximum MS response. | LC-MS/MS | View |
ACETEXL-101-1002U | Avantor® ACE® C18 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative. | LC-MS/MS | View |
ACETCORE-25A-0502U | Avantor® ACE® UltraCore™ SuperC18™ solid core (core shell) particle columns are ideal for demanding MS work offering improved MS signal intensity and maximum MS response. | LC-MS/MS | View |
ACETACE-111-1502 | Avantor® ACE® C18 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative. | LC | View |
ACETEXL-1111-1502U | Avantor® ACE® Excel® SuperC18™ columns provide high stability across an extended pH range. | LC | View |
ACETACE-1110-1546 | Avantor® ACE® C18-PFP phase utilises a specially developed ligand combining a C18 chain with integral PFP functionality. | LC | View |
ACETACE-121-2546 | Avantor® ACE® C18 columns provide great reproducibility and column lifetime. These columns are available in a wide range of particle sizes and dimensions from capillary to preparative. | LC | View |