QuantaBio ToughMix
Easily Amplify your Difficult Samples with Quantabio ToughMix. Get your free sample today! T&Cs apply.
- PerfeCTa® MultiPlex qPCR ToughMix®
- PerfeCTa® qPCR ToughMix®
- qScript™ 1-Step Virus ToughMix®
- qScript™ XLT One-Step RT-qPCR ToughMix®
- repliQa HiFi ToughMix
- AccuStart™ II PCR ToughMix®
- Ultra-Plex™ 1-Step ToughMix® (4X)
- AccuStart™ Genotyping ToughMix®
- Genomic DNA extraction from dried blood spots, Extracta™ DBS
Be it samples of DNA from plant, blood, soil, groundwater, wastewater, overcome your challenging samples with Quantabio ToughMix PCR reagents.
PCR inhibitors are present in crude samples, shutting down PCR amplification or quenching fluoroscent signals. Problematic inhibitors, such as polysaccharides, heme/hemoglobin, humic acid, melanin etc, are neutralized by Quantabio ToughMixes, helping to avoid expensive, time-consuming DNA purification or RNA purification steps for crude sample analysis
Tough-tested for countless applications!
Successful ToughMix PCR Amplification from samples such as:
- Microbial DNA from soil samples
- Genomic DNA from plant
- Pathogenic and viral DNA from wastewater
- Microbial DNA from groundwater
- DNA from environmental samples
- DNA from fresh blood samples
- DNA from dried blood samples
- Microbial DNA of food products
Avoid harsh DNA purification and RNA purification strategies and reduce RNA and DNA degradation in your crude samples
Don't just take our word for it, see what scientists & lab technologists have to say about Quantabio ToughMixes!
- "The simple and elegant way to unlock genetic information."
~Mei Baker, MD, FACMG, Co-Director of Newborn Screening - Wisconsin, Extracta DBA. - "Higher sensitivity, positives at lower Cq. Great results with plant DNA with high inhibition level."
~Paula S, Lab Coordinator INIAV, PerfeCTa qPCR ToughMix"
*Terms and Conditions:
- Promotion valid from now till 30th June 2023
- The free sampling program is only for smallest size pack. Subject to approval.
- VWR standard Terms and Conditions apply